Imprimer
Tissue grinder, Potter-Elvehjem, ground glass, KIMBLE®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SP885500-0019 SP885500-0021 SP885500-0022 SP885500-0023 SP885500-0024
SCERSP885500-0019EA 363 EUR
SCERSP885500-0019 SCERSP885500-0021 SCERSP885500-0022 SCERSP885500-0023 SCERSP885500-0024
Tissue grinder, Potter-Elvehjem, ground glass, KIMBLE®
Homogénéisateurs Broyeurs pour tissus
Potter-Elvehjem with all-glass construction designed for a motor drive using a stirrer adapter.
  • Replacement components are available and interchangeable
  • Clearance between pestles and tubes is 0,004 to 0,006"
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.