Imprimer
di-Ammonium hydrogénoorthophosphate P₂O₅ ≥ 53%, tech.
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): Ammonium phosphate dibasique, Ammonium phosphate diammonium
10301.A1 10301.A7
10301.A1EA 67.2 EUR
10301.A1 10301.A7
di-Ammonium hydrogénoorthophosphate P₂O₅ ≥ 53%, tech.
di-Ammonium hydrogénoorthophosphate
CAS No.: 7783-28-0 Molecular Formula: (NH4)2HPO4 Formula Weight: 132.06 Physical Form: -100 Mesh Powder Density: 1.619 Appearance: White Melting Point: 155° dec. Solubility: Freely soluble in water. Insoluble in alcohol, acetone Merck Reference: 14,542 MDL No.: MFCD00010891
Corrosion inhibitors, in fermentation, as flux for soldering tin, copper, brass, and zinc
Formule: (NH₄)₂HPO₄
Poids moléculaire: 132.06 g/mol
Point de fusion: 155 °C (déc.)
Densité: 1.619
Température de stockage: Température ambiante
Numéro MDL: MFCD00010891
Numéro CAS: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Indice Merck: 14,542