Imprimer
Detectable ballpoint pens, Economy, Hygostar®
Fournisseur: MENSCH FRANZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
85564 85565
MENS85564EA 118 EUR
MENS85564 MENS85565
Detectable ballpoint pens, Economy, Hygostar®
Stylos Stylos à encre
Hygostar® detectable ballpoint pens are available in blue, black and red. The colour of the case corresponds to the ink colour.
  • With fastening clip
  • Retractable pen point
  • Very robust even if dropped
  • Can write at very high and very low temperatures
Made of a smooth plastic body, ideal for food production and oily or dusty environments.
Certifications: HACCP compliant.