Iodure d'aluminium ≥95%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Synonyme(s):
Aluminium iodure
Purity: 95%
CAS No.: 7784-23-8
Molecular Formula: AlI3
Formula Weight: 407.69
Physical Form: Chunks
Density: 3.98
Appearance: White to brownish-purple
Odour: Pungent
Melting Point: 191°
Boiling Point: 360°
Solubility: Soluble in alcohol, ether, carbon disulfide
Merck Reference: 14,346
UN No.: 3260
MDL No.: MFCD00003427
Catalyst in organic synthesis
Formule:
AlI₃ Poids moléculaire: 407.7 g/mol Point d'ébullition: 360 °C Point de fusion: 191 Densité: 3.98 Température de stockage: Température ambiante |
Numéro MDL:
MFCD00003427 Numéro CAS: 7784-23-8 EINECS: 232-054-8 UN: 3260 ADR: 8 II Indice Merck: 12,00358 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...