Calcium carbonate ≥98%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
Carbonate de calcium
Purity: 98%
CAS No.: 471-34-1
Molecular Formula: CaCO3
Formula Weight: 100.09
Physical Form: Powder
Density: 2.930
Refractive Index: 1.6583
Appearance: White
Odour: Odourless
Melting Point: 800° dec.
Solubility: Practically insoluble in water. Soluble in diluted acids
Merck Reference: 14,1657
MDL No.: MFCD00010906
Manufacture of paint, rubber, plastics, etc. In food, cosmetics, and pharmaceuticals. As soil conditioner, neutralizer of surface waters, and as an industrial acid neutralizer
Formule:
CaCO₃ Poids moléculaire: 100.09 g/mol Point de fusion: 800 °C (déc.) Densité: 2.930 Température de stockage: Température ambiante |
Numéro MDL:
MFCD00010906 Numéro CAS: 471-34-1 EINECS: 207-439-9 Indice Merck: 14,1657 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...