Sulfate de caesium ≥99.9% (base métaux)
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
|
Purity: 99.9%
CAS No.: 10294-54-9
Molecular Formula: Cs2SO4
Formula Weight: 361.87
Physical Form: -20 Mesh Powder
Density: 4.243
Melting Point: 1010°
Solubility: Very soluble in water. Slightly soluble in alcohol, acetone, pyridine
Merck Reference: 14,2016
MDL No.: MFCD00010959
Formule:
Cs₂O₄S Poids moléculaire: 361.87 g/mol Point d'ébullition: 1627 Point de fusion: 1010 Densité: 4.243 Température de stockage: Température ambiante |
Numéro MDL:
MFCD00010959 Numéro CAS: 10294-54-9 EINECS: 233-662-6 Indice Merck: 14,2016 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...