Imprimer

Acide 4-acétamidobenzoïque 99+%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

158430250 158431000
ACRO158430250EA 58.1 EUR
ACRO158430250 ACRO158431000
Acide 4-acétamidobenzoïque 99+%
Acide 4-acétamidobenzoïque
Formule: C₉H₉NO₃
Point d'ébullition: 399 °C (1013 hPa)
Point de fusion: 255…262 °C
Numéro MDL: MFCD00002534
Numéro CAS: 556-08-1
EINECS: 209-114-7

Commander maintenant

Résultats des tests de spécification

Appearance Off-white to light beige Powder
Infrared spectrum Conforms
Melting point 255 to 262 °C
HPLC ≥99.0 %
Titration with NaOH 99.0 to 101.0 %
Sulfate (SO4) ≤0.02 %
AsO4, PO4, SiO4 (as SiO2) ≤0.001 %
Ammonium (NH4) ≤0.002 %
Heavy metals (as Pb) ≤0.005 %
Insoluble matter ≤0.01 % (In dilute NH4OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR