Imprimer
Baths for immersion thermostat, Termotronic II
Fournisseur: SELECTA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6000391 6000393 6000390 6000392
SELE6000391EA 471 EUR
SELE6000391 SELE6000393 SELE6000390 SELE6000392
Baths for immersion thermostat, Termotronic II
Bains Bains-marie
These metal baths are made up of double external AISI 304 stainless steel skin and an AISI 310 stainless steel pressed bath interior.