Imprimer

7,8-Dimethylalloxazine ≥98%, solide cristallin

Marques : Cayman Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): Lumichrome, 7,8-Dimethylbenzo[g]pteridine-2,4(1H,3H)-dione

20730-1 20730-100 20730-250 20730-500
CAYM20730-1EA 190 EUR
CAYM20730-1 CAYM20730-100 CAYM20730-250 CAYM20730-500
7,8-Dimethylalloxazine ≥98%, solide cristallin
7,8-Dimethylalloxazine

Lumichrome is a natural metabolite of riboflavin. It can be made by photolysis of riboflavin or it can be produced enzymatically in certain microbes and plants.


Lumichrome produced by Rhizobium and other bacterial species induces major developmental changes in plants at nanomolar concentrations. Lumichrome can stimulate larval metamorphosis in ascidians and activate the LasR quorum sensing receptor of bacteria. Lumichrome competitively inhibits the uptake of riboflavin by riboflavin transporters from prokaryotes and eukaryotes.


Lumichrome is sparingly soluble in methanol.


Synonym: NSC 96911; 7,8-dimethylbenzo[g]pteridine-2,4(1H,3H)dione

Formule: C₁₂H₁₀N₄O₂
Poids moléculaire: 242.24 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00005021
Numéro CAS: 1086-80-2
EINECS: 214-120-8

Commander maintenant

Résultats des tests de spécification

Purity ≥98%
λmax 219, 248, 335, 384 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR