Imprimer
Test tube racks for Immersion Thermostat
Fournisseur: SELECTA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1002531 1002532
SELE1002531EA 34.9 EUR
SELE1002531 SELE1002532
Test tube racks for Immersion Thermostat
Portoirs Tube Racks
These racks are designed for use with methacrylate tank.
  • Made of AISI 304 stainless steel
  • With rack linking guides