Imprimer
Horizontal wafer shippers, contactless, 200 mm SmartStack®
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HWS200C-4PK-CU HWS200C-1PK-KI HWS200C10161C0
ENTEHWS200C-4PK-CUEA 0 EUR
ENTEHWS200C-4PK-CU ENTEHWS200C-1PK-KI ENTEHWS200C10161C0
Horizontal wafer shippers, contactless, 200 mm SmartStack®
Matériel de manipulation de wafers Transport de semi-conducteurs
This SmartStack® contactless horizontal wafer shippers safely protect and transport bumped wafers in full, thin or ultrathin thickness.
  • Transports up to 25 wafers of varying thickness
  • Stackable configuration increases shipping density and is automation compatible
  • Used for thin, 3D, lensed or bumped wafers
Perimeter support ring minimizes the potential for surface area damage and eliminates the likelihood of scuffed, marred or bump damaged wafers. Rugged, engineering-grade polymer construction eliminates the need for TYVEK® separators and pink foam cushions.
Informations de commande: Shipper contains contactless horizontal wafer shipper and 26 rings. Shipper kit contains contactless horizontal wafer shipper, 26 rings, corrugated shipping box, outer packing cushions and CMB moisture barrier bag.