Imprimer
Test tubes, conical, with NS stopper
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2.588.001 2.588.002
WITG2.588.001EA 17.5 EUR
WITG2.588.001 WITG2.588.002
Test tubes, conical, with NS stopper
Tubes Tubes à essais
DURAN®, borosilicate glass, clear. With standard ground joint and PE stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes