Imprimer
Safety shoes, lace-up, jo_Max, Jori
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
12731/42 12731/43 12731/44 12731/45 12731/46 12731/47 12731/48 12731/38 12731/39 12731/40 12731/41
ELTN12731/42EA 69.9 EUR
ELTN12731/42 ELTN12731/43 ELTN12731/44 ELTN12731/45 ELTN12731/46 ELTN12731/47 ELTN12731/48 ELTN12731/38 ELTN12731/39 ELTN12731/40 ELTN12731/41
Safety shoes, lace-up, jo_Max, Jori
Chaussures et bottes
Antistatic safety shoe made from hydrophobised nubuck leather, mesh inserts and reflective materials, skin-friendly textile lining, a full, padded bellows tongue and collar, and a dual-density rubber/PU jo_MAX sole (3,0 mm profile, oil, fuel and chemical resistant, temperature resistant from approx. -20 and +200 °C).
  • Climate regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full-length inlay sole
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifications: EN ISO 20345; S3, SRC