Ethylènediamine 99%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Synonyme(s):
1,2-Ethanediamine, 1,2-Diaminoethane
Purity: 99%
CAS No.: 107-15-3
Molecular Formula: C2H8N2
Formula Weight: 60.10
Flash Point: 38°(100°F)
Physical Form: Liquid
Density: 0.899
Refractive Index: 1.4565
Sensitivity: Air Sensitive
Appearance: Colourless to pale yellow
Odour: Ammonia-like
Melting Point: 8-11°
Boiling Point: 117-118°
Solubility: Soluble in water, alcohol. Slightly soluble in ether
Beilstein Reference: 605263
Merck Reference: 14,3795
UN No.: 1604
MDL No.: MFCD00008204
Strongly basic amine useful as a building block in chemical synthesis
Formule:
C₂H₈N₂ Poids moléculaire: 60.1 g/mol Point d'ébullition: 117…118 °C Point de fusion: 8…11 °C Densité: 0.899 Point éclair: 38 °C (100 °F) Température de stockage: Température ambiante |
Numéro MDL:
MFCD00008204 Numéro CAS: 107-15-3 EINECS: 203-468-6 UN: 1604 ADR: 8 II Indice Merck: 13,03829 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...