Imprimer
Printer, Orion™ Star A series
Fournisseur: Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
STARA-106 STARA-108 STARA-109
ORIOSTARA-106EA 796 EUR
ORIOSTARA-106 ORIOSTARA-108 ORIOSTARA-109
Printer, Orion™ Star A series
Imprimantes Document Printers
This compact ink ribbon printer with built-in paper cutter can be used with Orion Star A210, Star A220 and Star A320 series meters; Dual Star meters; Versa Star and Versa Star Pro meters.
  • USB or Ethernet on-board interface
  • Serial, parallel, USB, Ethernet, Bluetooth and WiFi interfaces options
  • Cutter rated for 1 800 000 cuts
SDK Available for iOS and Android operating systems.
Certifications: FCC, RoHS, Energy Star certified.
Informations de livraison: Supplied with one ink ribbon, one paper roll, meter interface cable and power cords (110 V US/Japan, 220 V Euro, 240 V UK/Singapore and China/Australia/New Zealand).