Formiate de butyle ≥97%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Purity: 97%
CAS No.: 592-84-7
Molecular Formula: HCO2(CH2)3CH3
Formula Weight: 102.13
Flash Point: 18°(64°F)
Refractive Index: 1.3910
Melting Point: -92°
Boiling Point: 105-107°
Beilstein Reference: 1742108
UN No.: 1128
MDL No.: MFCD00003296
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...