Linoléate de méthyle 95%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
cis,cis-9,12-Octadecadienoic acid methyl ester, 9,12-Octadecanoic acid methyl ester, 9Z,12Z-Octadecadienoic acid methyl ester, Linoleic acid methyl ester, (9Z,12Z)-Methyl octadeca-9,12-dienoate, Methyl cis,cis-9,12-octadecadienoate, 9Z,12Z-Oktadekadiensäuremethylester, 9,12 Methyl octadecaotenoate methyl linoleate, cis,cis-9,12-Oktadekadiensäuremethylester
Purity: 95%
CAS No.: 112-63-0
Molecular Formula: C19H34O2
Formula Weight: 294.47
Storage Temperature: 2°C to 8°C
Flash Point: 200°(392°F)
Refractive Index: 1.4640
Melting Point: -35°
Boiling Point: 207-208°/11mm
Beilstein Reference: 1727614
Merck Reference: 14,6095
MDL No.: MFCD00009534
Formule:
C₁₉H₃₄O₂ Point d'ébullition: 207…208 °C (11 mmHg) Point de fusion: –35 °C Densité: 0,888 Point éclair: 200 °C (392 °F) |
Numéro MDL:
MFCD00009534 Numéro CAS: 112-63-0 EINECS: 203-993-0 Indice Merck: 14,6095 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...