Imprimer
ESD safety shoes, clogs, Michel
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
72430/40 72430/41 72430/42 72430/43 72430/44 72430/45 72430/46 72430/47 72430/48 72440/40 72440/41 72440/42 72440/43 72440/44 72440/45 72440/46 72440/47 72440/48
ELTN72430/40EA 57.9 EUR
ELTN72430/40 ELTN72430/41 ELTN72430/42 ELTN72430/43 ELTN72430/44 ELTN72430/45 ELTN72430/46 ELTN72430/47 ELTN72430/48 ELTN72440/40 ELTN72440/41 ELTN72440/42 ELTN72440/43 ELTN72440/44 ELTN72440/45 ELTN72440/46 ELTN72440/47 ELTN72440/48
ESD safety shoes, clogs, Michel
Chaussures et bottes
Lightweight anti-static clogs made from smooth, tear resistant microfibre upper, an adjustable Velcro® heel strap, and a mono-density PU sole (2,5 mm, oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole (C-Fit ESD)
  • ESD soft fleece insole
  • Shock-absorbent heel
  • Protective steel toecap (200 J)
Colour: Black or white
Certifications: EN 61340-5-1 (ESD); EN ISO 20345 SB, SRC, A, E, FO.