Potassium sodium L(+)-tartrate tetrahydraté ≥99%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
L-tartrate de sodium et de potassium tétrahydraté, Potassium-sodium L(+)-tartrate tétrahydraté, (R,R)-(+)-Tartrate de potassium et de sodium tétrahydraté
Purity: 99%
CAS No.: 6381-59-5
Molecular Formula: C4H4KNaO6.4H2O
Formula Weight: 282.23 (210.29anhy)
Physical Form: Crystals
Density: 1.77
Appearance: White
Odour: Odourless
pH: 6.0-8.5
Melting Point: 70-80°
Solubility: Soluble in water. Insoluble in alcohol
Beilstein Reference: 6113568
Merck Reference: 14,7670
Optical Rotation: +21° (c=1 in water)
MDL No.: MFCD00150989
Baking powder, medicine (cathartic), component of Fehling's solution, silvering mirrors
Formule:
KOOC(CHOH)₂COONa·4H₂O Point de fusion: 70…80 °C Densité: 1,77 Température de stockage: Température ambiante |
Numéro MDL:
MFCD00150989 Numéro CAS: 6381-59-5 EINECS: 206-156-8 Indice Merck: 14,7670 |