Imprimer
(S)-(-)-Acide malique ≥99%, en poudre
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
0210223790 0210223791 0210223725 0210223780
ICNA0210223790EA 207 EUR
ICNA0210223790 ICNA0210223791 ICNA0210223725 ICNA0210223780
(S)-(-)-Acide malique ≥99%, en poudre
(S)-(-)-Acide malique
L-Malic Acid is a dicarboxylic acid that is made by all living organisms. The malate anion is an intermediate in the citric acid cycle.
L-Malic Acid is a dicarboxylic acid that is made by all living organisms. The malate anion is an intermediate in the citric acid cycle.
Soluble in 100 mg/ml - clear to very slightly hazy, colourless.
Formule: C₄H₆O₅
Poids moléculaire: 134.09 g/mol
Point d'ébullition: 322
Point de fusion: 101…104 °C
Densité: 1.595 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00064213
Numéro CAS: 97-67-6
EINECS: 202-601-5
Indice Merck: 13,05730