Imprimer
Safety ankle boots, lace-up, Basic, Achim S3, Jori
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
16321/36 16321/37 16321/38 16321/39 16321/40 16321/41 16321/42 16321/43 16321/44 16321/45 16321/46 16321/47 16321/48
114-8376EA 41.9 EUR
114-8376 114-8377 114-8378 114-8379 114-8380 114-8381 114-8382 114-8383 114-8384 114-8385 114-8386 114-8387 114-8388
Safety ankle boots, lace-up, Basic, Achim S3, Jori
Chaussures et bottes
Durable safety ankle boots made from wear resistant cowhide leather, skin friendly textile lining, a full padded bellows tongue and collar padding plus a dual-density PU/PU New Basic sole (oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full length inlay sole
  • Anti-static soft fleece insole
  • PU scuff cap protects the upper material
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifications: EN ISO 20345; S3, SRC.