Imprimer
G418 solution_SC
Marques : Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): G-418, (2R,3S,4R,5R,6S)-5-Amino-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-2-((R)-1-hydroxyethyl)tetrahydro-2H-pyran-3,4-diol
L0015-020 L0015-100
L0015-020EA 82.7 EUR
L0015-020 L0015-100
G418 solution_SC
G418
Antibiotic G418 is an aminoglycoside antibiotic similar in structure to gentamycin. It exhibits toxicity towards both eukaryotic and prokaryotic cells. The optimal concentration for selection and maintenance must be determined for each cell line.
  • Clear to very slightly hazy liquid
  • Shelf life of 24 months
  • Storage temperature of –20 °C
Attention: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.
Formule: C₂₀H₄₀N₄O₁₀
Poids moléculaire: 496.552 g/mol
Numéro MDL: MFCD30541267
Numéro CAS: 49863-47-0