Imprimer
Penicillin/Streptomycin/Amphotericin B 100x
Marques : Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
L0010-100 L0010-020
L0010-100EA 43.9 EUR
L0010-100 L0010-020
Penicillin/Streptomycin/Amphotericin B 100x
Penicillin/Streptomycin/Amphotericin B
Antibiotic-antimycotic used in tissue culture to prevent growth of bacteria (gram positive and gram negative), yeast and fungi. Penicillin acts by inhibiting bacterial cell wall synthesis.
  • Yellow coloured, with a pH of 8,5±1
  • Shelf life of 24 month.
  • Endotoxin: <10 EU/ml
Formulation:
Sodium Chloride 8.50 g/L
Amphotericin B 0.025 g/L
Penicillin G Sodium Salt 6.03 g/L
Streptomycin Sulfate 10.00 g/L
Attention: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.