Imprimer
ESD safety shoes, sandals, uvex 1 ladies, 8562
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8560.7/38 8560.7/39 8560.7/40 8560.7/35 8560.7/41 8560.7/42 8560.7/43 8560.7/36 8560.7/37
UVEX8560.7/38EA 160 EUR
UVEX8560.7/38 UVEX8560.7/39 UVEX8560.7/40 UVEX8560.7/35 UVEX8560.7/41 UVEX8560.7/42 UVEX8560.7/43 UVEX8560.7/36 UVEX8560.7/37
ESD safety shoes, sandals, uvex 1 ladies, 8562
Chaussures et bottes
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable water resistant high-tech micro velour upper, soft padded tongue and collar, and a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Grey/pink
Width: 10
Certifications: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1, SRC