Imprimer
N,N-Diméthyl p-phénylènediammonium dichlorure ≥99.00% (par analyse titrimétrique) convient à l'analyse de la peroxydase, Sigma-Aldrich®
Marques : SIGMA ALDRICH MICROSCOPY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): N,N-Dimethyl-p-phenylenediamine dihydrochloride, N,N-Dimethyl-P-phenylenediamine 2HCl
M-Clarity™ (Niveau de Qualité) = MQ200
D4139-100G D4139-10G D4139-25G
D4139-100GEA 340 EUR
D4139-100G D4139-10G D4139-25G
N,N-Diméthyl p-phénylènediammonium dichlorure ≥99.00% (par analyse titrimétrique) convient à l'analyse de la peroxydase, Sigma-Aldrich®
N,N-Diméthyl p-phénylènediammonium dichlorure
N,N-Dimethyl-p-phenylenediamine dihydrochloride (DMPD) has been used in the HID-AB (high iron diamine-alcian blue) staining of intestinal tract samples. N,N-Dimethyl-p-phenylenediamine dihydrochloride (DMPD) is used for the determination of peroxidase. DMPD is converted into a red pigment of semiquinone nature. It is also used in the HID-AB (high iron diamine-alcian blue) staining procedures, which are used to differentiate between sialomucin and sulfomucin in the gastrointestinal tract.
Attention: Darkens readily when exposed to air.
Formule: (CH₃)₂NC₆H₄NH₂·2HCl
Poids moléculaire: 209.12 g/mol
Point de fusion: 222 °C (déc., lit.)
Température de stockage: Température ambiante
Numéro MDL: MFCD00012991
Numéro CAS: 536-46-9
EINECS: 208-635-7
UN: 2811
ADR: 6.1 II
Indice Merck: 13,03280
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f64621224792e0a4614d3b6a46f0a95e29b130a8&vid=f012289c4ede1e0a60734047c7b7b455f6eb867e&prid=web&clid=SAPDAM
Commander maintenant
Résultats des tests de spécification
Solubility (method) 50 mg/ml in water
Infrared spectrum Conforms to structure