Imprimer
Block heaters, analogue
Fournisseur: OHAUS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
30392047 30392054 30392068 30392075
OHAU30392047EA 494 EUR
OHAU30392047 OHAU30392054 OHAU30392068 OHAU30392075
Block heaters, analogue
Chauffe-blocs Bains à sec analogiques
These high wattage, constant-temperature dry block heaters are economical, versatile and compact. The multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures.
  • PID temperature controller maintains precise temperature control
  • Samples are heated to temperature quickly and accurately
  • Has the temperature uniformity of ±0,1 °C at 37 °C within the block
  • Can be used for coagulation and RH studies
Each model accepts separate interchangeable modular blocks, accommodating various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes. Each block has a thermometer well for measuring block temperature. Anodised aluminum modular blocks provide superior temperature stability and heat transfer.
Dual temperature control knobs with dial markings from 1 to 10 for low temperature and high temperature adjustments. Low range knob adjusts from ambient to 100 °C and high range knob adjusts from 75 to 150 °C.
Units can be run in conditions from 18 to 33 °C (64 to 91 °F), 20 to 80% relative humidity, non-condensing.
Informations de commande: Modular heating blocks must be ordered separately.
Informations de livraison: Units include a detachable, 3-wire cord and plug.
Attention: To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.