Imprimer
Acide nitrique 65 - 70% ≥99.999% (base métaux)
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): Nitric acid solution
87920.AE 87920.AP
87920.AEEA 88.9 EUR
87920.AE 87920.AP
Acide nitrique 65 - 70% ≥99.999% (base métaux)
Acide nitrique
Purity: 99.999% CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Density: 1.41 Appearance: Colourless to pale yellow Odour: Pungent Boiling Point: 121° Solubility: Very soluble in water and ether. Decomposes in alcohol Merck Reference: 14,6577 UN No.: 2031 MDL No.: MFCD00011349
In the manufacture of fertilizers and explosives, in organic synthesis, in metallurgy, in photoengraving, in etching steel, in rubber chemicals, and in reprocessing nuclear fuels
Formule: HNO₃
Point d'ébullition: 121 °C
Point de fusion: –42 °C
Densité: 1,41
Température de stockage: Température ambiante
Numéro MDL: MFCD00011349
Numéro CAS: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Indice Merck: 14,6577