Imprimer
Antigen retrieval reagent, IHC
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ADI9502710500 ADI9502720500 ADI9502730500
ENZOADI9502710500EA 136 EUR
ENZOADI9502710500 ENZOADI9502720500 ENZOADI9502730500
Antigen retrieval reagent, IHC
Protein Extraction Protein Extraction Reagents
Antigen retrieval reagent is used to recover antigens masked by fixation in cross linking fixatives such as formalin.