Imprimer

(±)-Acide bromosuccinique 96%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

B22829.06 B22829.14 B22829.22
B22829.06EA 36.6 EUR
B22829.06 B22829.14 B22829.22
(±)-Acide bromosuccinique 96%
(±)-Acide bromosuccinique

Purity: 96% CAS No.: 923-06-8 Molecular Formula: C4H5BrO4 Formula Weight: 196.99 Melting Point: 164-166° dec. Beilstein Reference: 1723556 Merck Reference: 14,1437 MDL No.: MFCD00004213

Formule: HOOCBrCHCH₂COOH
Point d'ébullition: 319 °C (1013 hPa)
Densité: 2,073 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00004213
Numéro CAS: 923-06-8
EINECS: 213-087-7
Indice Merck: 14,1437

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR