Imprimer
VWR®, DNA ladders
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
733-2602EA 50.8 EUR
733-2602 733-2601 733-2603
VWR®, DNA ladders
Réactifs d'électrophorèse Marqueurs de poids moléculaires
VWR® DNA ladders are convenient dsDNA ladders supplied in a loading buffer, which are ready to use on agarose and SDS DNA gels.
  • Ready to use loading buffer
  • For direct loading and easy visualisation
  • Suitable with TBE and TAE electrophoresis systems
  • Clear, distinct bands
  • Blue dye front running at 100 to 300 bp at 0,5 to 1,5% agarose
Informations de commande: Supplied in 0,5 ml packs for 100 lanes.