Imprimer
ARISTAR®VWR® , Cesium Standard Solution, 10000 mg/L Cs in dil. Nitric Acid (from CsNO₃), Standard for ICP
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
455212KEA 305 EUR
455212K 455214M
ARISTAR®VWR® , Cesium Standard Solution, 10000 mg/L Cs in dil. Nitric Acid (from CsNO₃), Standard for ICP
Standards Cationic Standards Cesium Standards
CsNO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.