Imprimer
VWR® ARISTAR®, Lead Standard Solution, 1000 mg/L Pb in dil. Nitric Acid (from Pb(NO₃)₂), Standard for ICP
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
455562GEA 200 EUR
455562G 455564Y
VWR® ARISTAR®, Lead Standard Solution, 1000 mg/L Pb in dil. Nitric Acid (from Pb(NO₃)₂), Standard for ICP
Standards Cationic Standards Lead Standards
Pb(NO₃)₂ in 2% HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.