Imprimer
Cleaning spray for electronics, KONTAKT WL
Fournisseur: HM INDUSTRIESERVICE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
100025
291-1246EA 20 EUR
291-1246
Cleaning spray for electronics, KONTAKT WL
Produits nettoyants Détergents et Nettoyants de Surfaces
KONTAKT WL is the universal spray wash for all electronics. The solvent mixture removes a wide range of dirt such as oil, grease, soot or nicotine residues from electrical devices, plugs, engines or precision mechanical gears.
KONTAKT WL has a high dissolving power, good material compatibility with common electronic components and dries quickly and without leaving any residue.