Imprimer
Precision heating bath
Fournisseur: GESTIGKEIT HARRY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PZ26-4 PZ26-7
GESTPZ26-4EA 2040 EUR
GESTPZ26-4 GESTPZ26-7
Precision heating bath
Bains Bains à ébulition
Precision heating baths which can be used for fluid, oil, air or as a sandbath for continuous operation. They are very well suited for use with rotary evaporators, heating is provided by a tubular radiator.
  • Made of stainless steel with spray-painted base
The appliances are controlled by means of a microprocessor PID-controller in connection with a low-mass sensor. The actual-value temperature is always on view. The preset temperature can be seen by pushing a button and can also be adjusted with the up/down key.