Imprimer
Drum spill kits
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
813869 813870 813871
BRDY813869EA 383 EUR
BRDY813869 BRDY813870 BRDY813871
Drum spill kits
Matériaux absorbants
The drum spill kit is designed for medium sized spills.
  • Water and chemical resistant drum protects sorbents
  • Protects sorbents against moisture, dirt and damage
  • Drum can also be used as shipping container for used sorbents
  • Easy access to products with screw top lid
Informations de livraison: Supplied with 12 pads (410×510 mm), 3 socs (Ø 7,6×3660 mm), 2 pillows (430×480 mm), 1 pair of gloves, 1 pair of goggles and 3 disposal bags.