Imprimer
Heat transfer liquids, Kryo 60
Marques : LAUDA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
LZB102 LZB202 LZB302
461-2853EA 800 EUR
461-2853 461-2859 461-2871
Heat transfer liquids, Kryo 60
Liquides pour bains à circulation
These transfer fluids allow highly accurate thermostating, even at extreme temperatures.
  • Durability
  • Simple and safe handling
  • Reliability, suitable for long-term operation
Recommended temperature range: –60 to +60 °C (open system).
* Denotes silicone oil