Imprimer
Burettes, straight stopcock, PTFE spindle, Schellbach, BLAUBRAND®
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
912488 912487 912484
BRND912488EA 159 EUR
BRND912488 BRND912487 BRND912484
Burettes, straight stopcock, PTFE spindle, Schellbach, BLAUBRAND®
Burettes
Borosilicate glass 3.3, class AS.
  • Calibrated to deliver 'TD, Ex', DE-M marking
  • Schellbach stripe, ring marks at major graduations
  • Marks and inscriptions in high contrast blue enamel represent the optimum combination of resistance and readability
Length: 800 mm
With individual certificate
Certifications: DIN EN ISO 385