Imprimer
Heat resistant welding gloves, ActivArmr® 43-217
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
43-217/10 43-217/11 43-217/9
ANSE43-217/10EA 103 EUR
ANSE43-217/10 ANSE43-217/11 ANSE43-217/9
Heat resistant welding gloves, ActivArmr® 43-217
Gants
This TIG welding glove made from full grain goatskin provides EN 12477 Type-B protection for precise welding and thermal tasks.
  • Provides protection from heat, flames, sparks and weld spatter
  • 3-dimensional thumb design greatly increases fine dexterity for precise welding control
  • Premium full grain goatskin and welted seams with para-aramid thread ensure durability in use
  • Long split cowhide cuff extends protection to the forearms
Suitable for TIG welding, as well as grinding, brazing, metal fabrication and maintenance.
Certifications: EN 407 (41XX4X), EN 12477 Type-B; Cat. III