Imprimer
TE buffer, 1X, Molecular Biology Grade, Ultrapure
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
J75893.XX J75893.AE J75893.AP
J75893.XXEA 64.6 EUR
J75893.XX J75893.AE J75893.AP
TE buffer, 1X, Molecular Biology Grade, Ultrapure
Tampons Tampons biologiques
Use directly to re-suspend and/or dilute purified DNA or RNA.
  • 1X TE solution consisting of 10 mM Tris, and 1 mM EDTA
  • Solution is 0,2 μm filtered and dispensed into vials and/or durable, square Nalgene bottles