Imprimer
pH buffer solutions, NIST/DIN
Marques : Mettler - Toledo
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
51350052 51350054 51350058 51350056 30111136 30111137 30111138 30111139
METT51350052EA 34.8 EUR
METT51350052 METT51350054 METT51350058 METT51350056 METT30111136 METT30111137 METT30111138 METT30111139
pH buffer solutions, NIST/DIN
Tampons Tampons pH
The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision.
A detailed test certificate is available for every bottle and guarantees the tested values and traceability.
Certifications: NIST traceable.