Imprimer
Cleaning cloths, MicroClean Plus
Fournisseur: FHP VILEDA SCS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
152534 152530 152533 152531 152532
VILE152534EA 572 EUR
VILE152534 VILE152530 VILE152533 VILE152531 VILE152532
Cleaning cloths, MicroClean Plus
Lingettes Chiffons nettoyants réutilisables Chiffons pour applications générales
MicroClean Plus is a high performance cleaning cloth made from a uniquely fine microfibre (70% polyester and 
30% polyamide). Great streak- and lint-free results are achieved with just one wipe. Ideal for quick dirt removal even without chemicals.
  • Extremly durable
  • Micron Fibre Technology for deeper and more effective cleaning
  • MicroClean dries quickly: Bacteria growth is prevented
  • Washable and reusable: Reduces waste and protects the environment
  • Colour coding system allows a cleaning process according to hygiene standards
MicroClean Plus is suitable for demanding cleaning tasks in general building cleaning and Hotels/Restaurants/Catering on every kind of surface - with or without cleaning agents.