Imprimer
Renal epithelial cell growth medium 2
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-26030 C-26230 C-26235 C-26130
C-26030EA 291 EUR
C-26030 C-26230 C-26235 C-26130
Renal epithelial cell growth medium 2
Milieux de culture cellulaire
The PromoCell Renal Epithelial Cell Growth Medium 2 is an optimised low-serum (5% V/V) medium with epinephrine. The combination of low serum and selected growth factors ensures successful growth in vitro and makes this medium the first choice for renal epithelial cells. The media are optimised for primary human cells but can also be used for porcine renal epithelial cells.
Recommended for Human Renal Epithelial Cells (HREpC) and Human Renal Cortical Epithelial Cells (HRCEpC).
Informations de commande: PromoCell Renal Epithelial Cell Media are available as a Medium (ready-to-use) or as a Medium Kit. The Medium (ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as SupplementMix and SupplementPacks can also be purchased separately.