Imprimer
Precolumn filters, ColumnSaver
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
55214-U 55215-U
55214-UEA 292 EUR
55214-U 55215-U
Precolumn filters, ColumnSaver
Filtres Filtres de précolonnes
Precolumn filter kit has a standard 10-32 fitting and fits direct to most column types without introducing an additional dead volume.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable