Imprimer
Capillary columns, traditional phases, SP-2380, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
24110-U 24111 24116-U 25319
24110-UEA 748 EUR
24110-U 24111. 24116-U 25319.
Capillary columns, traditional phases, SP-2380, Supelco®
Colonnes pour chromatographie
These columns are situated between the traditional non bonded cyanosilicone phases SP-2330 and SP-2340 in polarity. The high polarity of this phase allows the separation of geometric (cis/trans) fatty acid methyl ester isomers as a group.
  • Temperature limit from subambient to 275 °C
Certifications: Meets USP G48 requirements.