Imprimer
Bulb pipettes, BLAUBRAND® ETERNA, one mark, Class AS (one batch)
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2.30512 2.30507 2.30518
BRND2.30512EA 89.7 EUR
BRND2.30512 BRND2.30507 BRND2.30518
Bulb pipettes, BLAUBRAND® ETERNA, one mark, Class AS (one batch)
Pipettes Pipettes jaugées
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Including one batch certificate
  • Ring mark and inscriptions in ETERNA amber stain
  • Total order from one batch: All supplied pipettes come from one batch for statistical testing equipment monitoring
Certifications: DIN EN ISO 648; DE-M marking