Imprimer
3D tumorsphere medium (cancer stem cell medium)
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-28070 C-28075
C-28070EA 220 EUR
C-28070 C-28075
3D tumorsphere medium (cancer stem cell medium)
Milieux de culture cellulaire
PromoCell 3D Tumorsphere Medium has been designed to meet your requirements for the extended serial 3D tumorsphere culture. 3D Tumorsphere Medium supports the tumorsphere/mammosphere culture of the most commonly used cancer cell lines.
  • Directly switch from 2D to 3D culture
  • Supports most commonly used cancer cell lines
  • Allows extended serial tumorsphere passages (>10)
  • High tumorsphere formation efficiency
  • Xeno-free and serum-free defined formulation
  • Ready to use
Cell types tested for serial passage with the PromoCell 3D Tumorsphere Medium: Brain U-87 MG, Breast MCF-7; Colon HT-29, Connective tissue HT1080; Liver HepG2; Lung A-549; Pancreas Panc-1; Prostate LNCaP; Skin A-431.
The PromoCell Cancer Stem Cell Medium is ready-to-use and chemically defined providing a culture environment devoid of all stimuli originating from non-defined materials. It is particularly suitable as a cost-efficient tool for the standardized routine-culture of tumorspheres/mammospheres.
List of cell types tested for serial passage with the PromoCell: Bran U-87 MG, Breast MCF-7; Colon HT-29, Connective tissue HT1080; Liver HepG2; Lung A-549; Pancreas Panc-1; Prostate LNCaP; Skin A-431
Informations de commande: PromoCell 3D Tumorsphere Medium (Cancer Stem Cell Medium) is available as Medium (ready to use) and consists of a bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete Medium.