Imprimer
Drums, UV safe, with screw lids
Fournisseur: CURTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6942-91-101 7006-61-101 7015-61-101 7026-61-101
CURT6942-91-101EA 157 EUR
CURT6942-91-101 216-5000 CURT7015-61-101 CURT7026-61-101
Drums, UV safe, with screw lids
Barrels, Kegs, Drums
Drums, made of UV safe HDPE, with screw lid. For storage and shipment of UV sensitive powders, pastes and liquids.
  • Water tight
  • Tamper evident
  • Stackable
  • UN marking (1H2) for hazardous solids
  • Food grade