Imprimer
Conductivity cells, IDS
Fournisseur: WTW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
301711 301710 301720 301716 301721 301718 301719 301722
663-0153EA 694 EUR
663-0153 663-0152 663-0156 665-0304 665-0475 665-0408 665-0474 WTWA301722
Conductivity cells, IDS
Électrodes Électrodes de conductivité
High quality, enhanced sensor technology combined with most up to date measurement electronics. For use with IDS meters.
  • Intelligent: Sensors are automatically recognised by the meter, each sensor is individually identified
  • Digital: Measurement signals are processed digitally, providing very accurate data
  • IDS cells store their serial number and calibration records
  • TetraCon® 925: 4-electrode measuring cell ideal for medium and high conductivity applications
  • LR 925/01: Ideal for pure water and low conductivity applications
  • TetraCon® 925/LV: Graphite 4-electrode cell for small volumes and viscous samples
The electrodes can provide the following data, shown on the screen of the digital meter:
- Model name
- Serial number
- Calibration status (+++, ++, +, -, blank)
- The calibration history includes the last 10 calibrations, including buffers used, mV values, etc.
Informations de commande: Diverses longueurs de câble disponibles.