Imprimer
pH indicator, bromo cresol purple tablets, Lovibond®
Marques : Lovibond
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
515700BT 515701BT
LOVI515700BTEA 49.2 EUR
LOVI515700BT LOVI515701BT
pH indicator, bromo cresol purple tablets, Lovibond®
Kits de tests environnementaux
Indicator for pH range 5,2 to 6,8.
Can be used for measuring pH in conjunction with the Comparator 2000+ system.