Imprimer
Trypsin neutralizing solution
Marques : PROMOCELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-41100 C-41110 C-41120
C-41100EA 44 EUR
C-41100 C-41110 C-41120
Trypsin neutralizing solution
Suppléments et additifs pour culture cellulaire
Trypsin is the most widely employed enzyme used for cell detachment. This solution exhibit standardized enzyme activity and contain trypsin from extra-pure lots. This solution contains 0,05% trypsin inhibitor in 0,1% BSA.
Attention: For in vitro research use only and not for diagnostic or therapeutic procedures.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Species
Source
Learn more